Order Entry
ContactUsLinkComponent
TEST KIT CHLORINE HIGH CN-21P
Chlorine (Total) Test Kit, Model CN-21P, Hach
Catalog #: CA24444-00
Supplier:  Hach
CAS Number:  
TEST KIT CHLORINE HIGH CN-21P
Chlorine (Total) Test Kit, Model CN-21P, Hach
Catalog #: CA24444-00
Supplier:  Hach
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Chemical method:
    Thiosulfate
  • Description:
    Chlorine (Total) Test Kit, Model CN-21P
  • Model:
    CN-21P
  • Method:
    Drop count titration/ Thiosulfate
  • Case style:
    B
  • Cat. no.:
    CA24444-00
  • Range:
    10 - 200 mg/l
  • Supplier No.:
    24444-00
  • No. of Tests:
    100

Specifications

About this item

Chlorine Test Kits are manufactured to ensure compliance with stringent quality control standards.

This product has not been evaluated to test for chlorine and chloramines in medical applications in the United States.

  • Simple, economical titration method
  • Contains all required reagents, solutions, and glassware

Using the Drop Count Titration method, the CN-21P Chlorine Test Kit is used to determine high concentrations of chlorine in sanitizing applications

This product has not been evaluated to test for chlorine and chloramines in medical applications in the United States.

10-200 mg/L.

Caution: Hazardous Items: Items with this mark may be considered hazardous under some shipping conditions. If necessary, we will change your selected shipping method to accomodate these items.

**mg/L unless otherwise noted, ppb = μg/L, ppm = mg/L; gpg = grains per gallon; 1 gpg = 17.1 mg/L or 17.1 ppm. 1EPA-accepted for wastewater. Check with your regulatory agency.
Customers Who Bought This Also Bought