Sample Cells, Round Glass, Hach
Catalog #: CA24019-06
Supplier: Hach
Specifications
- Capacity:25 ml
- Ø:25 mm
- Description:Round Glass Sample Cells with 10, 20 and 25 mL Graduations
- Cat. no.:CA24019-06
- Supplier No.:24019-06
Specifications
About this item
Round, glass sample cells for use with Hach Spectrophotometers
Cells come in pack of six with caps.
Customers Who Bought This Also Bought