COD Standard Solutions, NIST, Hach
Catalog #: CA22539-29
Supplier: Hach
CAS Number:
Specifications
- Concentration:1000 mg/L as COD
- Packaging:Bottle
- Description:COD Standard Solution
- Size:200 ml
- Supplier No.:22539-29
Specifications
About this item
For accuracy checks in Chemical Oxygen Demand (COD) determinations.
Customers Who Bought This Also Bought