KIMTECH SCIENCE® Precision Wipes Tissue Wipers, KIMBERLY-CLARK PROFESSIONAL®
Catalog # CA21903-025
Supplier: Kimberly-Clark
Specifications
- Cat. no.:CA21903-025
- Packaging:280/Box
- W×L:11.2 x 21.3 cm (413/32 x 813/32")
- Supplier No.:05511
- Ply:1
Specifications
About this item
Soft, absorbent tissue wipers are an economical opiton that offers reliable wiping performance for delicate tasks. Coated POP-UP™ box helps protect wipers from spills.
- Antistatic, economical wipers
- Coated POP-UP® box protects wipes
- Color: White
For use in laboratories.
Frequently Bought Together