ContactUsLinkComponent
SpoKlenz* Ready-To-Use Sterilant Disinfectant
Spor-Klenz® Ready-To-Use Sterilant/Disinfectant, STERIS®
Catalog #: CA21899-864
Supplier:  Steris
SpoKlenz* Ready-To-Use Sterilant Disinfectant
Spor-Klenz® Ready-To-Use Sterilant/Disinfectant, STERIS®
Catalog #: CA21899-864
Supplier:  Steris
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    Spor-Klenz® Ready-to-Use Sterilant
  • Size:
    3.2 L (0.85 gal.)
  • Packaging:
    Pour Bottle
  • Supplier No.:
    652501CA
Specifications
About this item
Spor-Klenz® fast-acting, liquid cold sterilant/disinfectant requires no mixing or activation
Spor-Klenz® fast-acting, liquid cold sterilant/disinfectant requires no mixing or activation
Use wherever critically high-level microbial control is required. Ready-to-use format eliminates errors in dilution preparation. Low-toxicity profile adds to worker safety. Undiluted, the product is sporicidal in 51/2 hours and disinfects in 10 minutes. Accompanied by certificates of analysis, filtration, and sterility.
A fast acting, liquid cold sterilant/disinfectant, filtered to 0.22 micron and specifically formulated for use in the sterilization and disinfection of hard environmental surfaces in Pharmaceutical, Medical Device, Biotech and Cosmetic manufacturing facilities. This product is a stabilized blend of peracetic acid, hydrogen peroxide, and acetic that provides fast, effective microbial control, including spores.
Customers Who Bought This Also Bought