Order Entry
ContactUsLinkComponent
NESSLERS EX ALK MDB RG 100ML
Nessler Reagent, 100 ml, Hach
Catalog #: CA21194-32
Supplier:  Hach
CAS Number:  
NESSLERS EX ALK MDB RG   100ML
Nessler Reagent, 100 ml, Hach
Catalog #: CA21194-32
Supplier:  Hach
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Chemical method:
    Nessler's reagent
  • Description:
    Nessler Reagent
  • Size:
    100 mL MDB
  • Method:
    Nessler
  • Cat. no.:
    CA21194-32
  • Range:
    0.02-2.50 mg/L NH3-N
  • Supplier No.:
    21194-32

Specifications

About this item

For ammonia nitrogen determination by the Nessler method (Hach method 8038)

EPA accepted for wastewater analysis (distillation required). Range: 0.02-2.50 mg/L NH3-N. Also used in visual test kits - alternate ranges apply.

Ordering information: Contains mercury. Dispose of according to local, state and federal regulations.

Hazardous Items: Items with this mark may be considered hazardous under some shipping conditions. If necessary, we will change your selected shipping method to accommodate these items.
Customers Who Bought This Also Bought