Order Entry
ContactUsLinkComponent
Cell Strainers
Falcon® Cell Strainers, Sterile
Catalog #: CA21008-952
Supplier:  Corning
Cell Strainers
Falcon® Cell Strainers, Sterile
Catalog #: CA21008-952
Supplier:  Corning

Specifications

  • Pore size:
    70 µm
  • Color:
    White
  • Supplier No.:
    352350

Specifications

About this item

Falcon® cell strainers are an easy, ready-to-use way to consistently obtain a more uniform single-cell suspension. These cell strainers are made of a strong nylon mesh and are available with 40, 70, or 100 µm pores that are evenly spaced for optimal performance in a variety of applications. A faster and easier alternative to gauze filtration in procedures involving dissociation of cells from either clumps or primary tissue.

  • Moulded, color-coded PP frame with tab enables easy handling
  • Fits perfectly into a 50 ml Falcon® conical tube or other similarly sized tube
  • Sterilised by gamma irradiation and conveniently available in individual, peel-open packaging
  • Extended lip on strainer enables aseptic handling with forceps
Ordering information: Packaged individually in bags.
Customers Who Bought This Also Bought