Specifications
- Lens Coating:Uvextreme®
- Lens Tint:Clear
- Description:Ultra-spec 1000 glasses
- Cat. no.:CA13500-884
- Supplier No.:S0250X
Specifications
About this item
Economical glasses feature a 100% polycarbonate wraparound unilens design.
Economical glasses feature a 100% polycarbonate wraparound unilens design. Side shields add extra protection and provide optimal coverage without compromising visibility. Uvex Ultra-spec® 2000 glasses feature an adjustable temple length. Uvex Ultra-spec® 2001 OTG glasses fit over prescription lenses and feature adjustable Duoflex™ temples with soft, behind-the-ear comfort cushions. Clear lenses and frames
The Ultra-dura® lens coating provides superior scratch and chemical resistance. The Uvextreme® lens coating provides antifog, anti-scratch, antistatic, and anti-UV protection.
Lenses meet ANSI Z87+ and CSA Z94.3 specifications.
- Features 100% polycarbonate wraparound unilens design
- Side shields add extra protection without compromising visibility
- Clear lenses and frames
- The Ultra-dura® lens coating provides superior scratch and chemical resistance
- Uvextreme® lens coating provides anti-fog, anti-scratch, anti-static, and ultaviolet protection
Uvex Ultra-spec® 2000 glasses feature an adjustable temple length. Uvex Ultra-spec® 2001 OTG glasses fit over prescription lenses and feature adjustable Duoflex™ temples with soft, behind-the-ear comfort cushions. Clear lenses and frames.
The Ultra-dura® lens coating provides superior scratch and chemical resistance. The Uvextreme® lens coating provides antifog, anti-scratch, antistatic, and anti-UV protection.
Side shields add extra protection and provide optimal coverage without compromising visibility.