Convertors® Paper/Film Gas/Steam Sterilization Pouches, Cardinal Health®
CA11213-044
- Cat. no.:CA11213-044
- W×L:457×559 mm (18×22")
- Supplier No.:91822
Peel-open pouches are designed for quick, easy presentation of sterile items.
- Can be used in steam or EO sterilization
- Chemical process indicators on every pouch identify the sterilization process used
Heat-seal pouches can be heat-sealed or taped shut. Self-seal pouches feature a pressure-sensitive closure that is prefolded to help ensure the integrity of the seal.
Items within the pouch are easily viewed through the blue-tinted 2 mil-thick film side. The other side is made of tough, moisture-resistant, surgical-grade paper, which provides high porosity for fast sterilization while maintaining an effective bacterial barrier.
Frequently Bought Together