ContactUsLinkComponent
V. Mueller* DualPeel* Self-Sealing Pouch
Convertors® Paper/Film Gas/Steam Sterilization Pouches, Cardinal Health®
Catalog #: CA11206-415
Supplier:  Cardinal Health
Convertors® Paper/Film Gas/Steam Sterilization Pouches, Cardinal Health®
Catalog #: CA11206-415
Supplier:  Cardinal Health
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
  • W×L:
    89×222 mm (3.5×8¾")
  • Cat. no.:
    CA11206-415
  • Supplier No.:
    92308
Specifications
About this item
Peel-open pouches are designed for quick, easy presentation of sterile items.
  • Can be used in steam or EO sterilization
  • Chemical process indicators on every pouch identify the sterilization process used
Heat-seal pouches can be heat-sealed or taped shut. Self-seal pouches feature a pressure-sensitive closure that is prefolded to help ensure the integrity of the seal.
Items within the pouch are easily viewed through the blue-tinted 2 mil-thick film side. The other side is made of tough, moisture-resistant, surgical-grade paper, which provides high porosity for fast sterilization while maintaining an effective bacterial barrier.
Customers Who Bought This Also Bought