ContactUsLinkComponent
MycoAlert Mycoplasma Detection Kit
MycoAlert™ Mycoplasma Detection Kits, Lonza
Catalog #: CA11006-554
CAS Number:  
MycoAlert Mycoplasma Detection Kit
MycoAlert™ Mycoplasma Detection Kits, Lonza
Catalog #: CA11006-554
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    MycoAlert™ Mycoplasma Detection Kits
  • Cat. no.:
    CA11006-554
  • Supplier No.:
    LT07-318
  • No. of Tests:
    100
Specifications
About this item
The MycoAlert™ PLUS Mycoplasma Detection Kit is the next generation Lonza MycoAlert™ Kit. It provides higher light output to allow for use with less sensitive plate luminometers or multifunctional readers and testing of unused media, media supplements or water.
  • Convenient & fast
  • Simply add two reagents to the culture supernatant, perform two luminescence readings and get your results in less than 20 minutes
  • Detects all common mollicute contaminations (Mycoplasma, Acholeplasma, Entomoplasma and Spiroplasma; except Ureaplasma)
  • Suited for tube luminometers, plate luminometers and multifunctional readers
The MycoAlert™ PLUS Assay is a selective biochemical test that exploits the activity of mycoplasmal enzymes which are found in all six of the main mycoplasma cell culture contaminants and the vast majority of 180 mycoplasma species, but are not present in eukaryotic cells
MycoAlert™ Assay Control Set (available separately)
The MycoAlert™ Assay Control Set includes a lyophilized positive control and assay buffer for reconstitution. The assay buffer also serves as a negative control. The positive and negative controls are designed for use with the MycoAlert™ PLUS Mycoplasma Detection Kit. It is recommended to always include the Control Set to ensure that the operator and all of the assay reagents are performing properly. The positive control is proprietary, but does NOT contain mycoplasma. The positive or negative control is simply substituted for a sample in the standard assay protocol. The positive control will give a ratio of >1 while the negative control will give a ratio of <1.
Viable mycoplasma in a test sample (cell supernatant or fresh media or supplements) are lysed and the enzymes react with the MycoAlert™ PLUS Substrate, catalyzing the conversion of ADP to ATP. The ATP is then transferred into a light signal via the luciferase enzyme in the MycoAlert™ PLUS Reagent. By measuring the level of ATP in a sample both before (read A) and after the addition of the MycoAlert™ PLUS Substrate (read B), a ratio can be obtained which is indicative of the presence or absence of mycoplasma.
Customers Who Bought This Also Bought