ContactUsLinkComponent
GLUCOSE DRINK ORANGE 50G CS24
Glucose Tolerance Drinks, Azer Scientific
Catalog #: CA11006-153
Supplier:  Azer Scientific
GLUCOSE DRINK  ORANGE 50G CS24
Glucose Tolerance Drinks, Azer Scientific
Catalog #: CA11006-153
Supplier:  Azer Scientific
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    Glucose tolerance drink, orange, glucose concentration 50 g
  • Cat. no.:
    CA11006-153
  • Supplier No.:
    10-0-050-CA
Specifications
About this item
FDA-approved, non-carbonated beverages are designed for use in conjunction with glucose tolerance tests.
  • Shatterproof Plastic Bottles
  • Graduated Labels on Bottles
  • Non-Carbonated Beverages
  • Citric Acid Free
  • Dairy and Gluten-Free
All beverages are supplied in shatterproof plastic bottles.
Beverages are gluten- and dairy-free, certified Kosher, and are manufactured according to WHO and ADA standards. Simply Pure unflavored beverage is also citric acid free. The integrity of each batch is certified by an FDA recognized independent testing laboratory. All bottles are marked with an expiration date determined via accelerated and real time stability studies.
A graduated label on each bottle indicates the amount dispensed.
Certifications: Beverages are gluten and dairy free, certified Kosher, and are manufactured according to WHO and ADA standards. Simply Pure unflavored beverage is also citric acid free. The integrity of each batch is certified by a FDA-recognized independent testing laboratory. All bottles are marked with an expiration date determined via accelerated and real-time stability studies.
Ordering information: Beverages are packaged in four shrink-wrapped six-packs per case. Beverages are packaged in four shrink-wrapped six-packs per case.
Customers Who Bought This Also Bought