KIMTECH PURE® M6 Face Mask, Kimberly-Clark Professional®
Catalog #: CA10843-001
Supplier: Kimberly-Clark
Specifications
- Description:KIMTECH PURE* M6 face mask
- Cat. no.:CA10843-001
- Supplier No.:62477
Specifications
About this item
Two-layer general-purpose mask with earloops provides comfort with BICOSOF® fabric inner facing.
- Two layer, general purpose
- Earloops
- BICOSOF® fabric inner facing
Recommended for ISO Class 6 or higher cleanroom environments.
Packaging: Double-bagged.
Customers Who Bought This Also Bought