Order Entry
ContactUsLinkComponent
PerfeCTa Sybr Smx, Rox, 500R
PerfeCTa® SYBR® Green SuperMix Reaction Mixes, Quantabio
Catalog #: CA101414-158
Supplier:  Quantabio
CAS Number:  
PerfeCTa Sybr Smx, Rox, 500R
PerfeCTa® SYBR® Green SuperMix Reaction Mixes, Quantabio
Catalog #: CA101414-158
Supplier:  Quantabio
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Application:
    qPCR dye
  • Cycler compatibility:
    ABI 7900HT/7900HT Fast/StepOnePlus™
  • Description:
    PerfeCTa® SYBR® Green SuperMix, ROX™
  • Cat. no.:
    CA101414-158
  • No. of Reactions:
    500
  • Supplier No.:
    95055-500

Specifications

About this item

Unique buffer and stabilizers have been optimized exclusively for SYBR® Green I qPCR to deliver maximum PCR efficiency, sensitivity, and a robust fluorescent signal

  • 2X concentrated reagents minimize pipetting steps and improve accuracy
  • Ultrapure, antibody hot start AccuStart enzyme technology and anti-foaming formulation
  • Maximum dye concentration for robust optical signal with small amplicons (i.e. microRNAtemplated cDNA)

Kits are available for all real-time PCR instrument platforms including those requiring normalization with ROX™ reference dye. Platforms not requiring passive reference dye include Roche LightCycler® 400, Bio-Rad® iQ™, MyiQ™, iQ™5, Opticon®, and Chromo4™, Corbett Rotor-Gene®, Eppendorf® Mastercycler®, and Cepheid® SmartCycler®. Kits with Low ROX™ reference dye are for use with platforms including AB 7500 and Stratagene® Mx™. Kits with ROX™ reference dye are for use with platforms including AB 7000, 7300, 7700, 7900, and StepOne™.

All SuperMixes are available with or without uracil-N-glycosylase (UNG) for prevention of carry-over contamination from previous dU containing PCRs.

Highly specific amplification is crucial to successful qPCR with SYBR® Green I technology because this dye binds to and detects any dsDNA generated during amplification. A key component of this supermix is AccuStart™ Taq DNA polymerase with monoclonal antibodies that bind to the polymerase and keep it inactive prior to the initial PCR denaturation step. Upon heat activation (2 minutes at 95°C), the antibodies denature irreversibly, releasing fully active, unmodified Taq DNA polymerase. This enables specific and efficient primer extension with the convenience of room temperature reaction assembly.

Customers Who Bought This Also Bought