Order Entry
ContactUsLinkComponent
ORION IONPLS SILVR/SULF ELE BN
Orion™ Silver/Sulfide Electrodes, Thermo Scientific
Catalog #: CA10010-810
ORION IONPLS SILVR/SULF ELE BN
Orion™ Silver/Sulfide Electrodes, Thermo Scientific
Catalog #: CA10010-810
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Max. I.D.:
    0 to 80°C
  • Temperature range:
    0 to 80°C
  • Description:
    Ionplus Sure-Flow Solid state combination silver/sulfide electrode, with waterproof BNC connector
  • Cat. no.:
    CA10010-810
  • Cable Length:
    1 m
  • Supplier No.:
    9616BNWP

Specifications

About this item

Make quick, simple, accurate, and economical measurements of silver and sulfide ions in aqueous solutions.

  • Reference and sensing electrodes are built into one electrode
  • Sure-Flow reference junction prevents electrode clogging and provides fast and stable readings
  • Can be used on any ISE meter with a BNC connection
  • Free-flowing liquid junction assures stable, drift-free potentials

Orion™ Silver/Sulfide Electrodes have dual electrode measuring silver and sulfide ions in aqueous solutions and perform low-level cyanide and halide titrations.

Customers Who Bought This Also Bought