To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Description:Combination sodium electrode, with waterproof BNC connector
- Cat. no.:CA10010-776
- Measurement Range:Saturated to 10⁻⁶ M; saturated to 0.02 ppm
- Cable Length:1 m
- Supplier No.:8611BNWP
Specifications
About this item
Measure sodium in aqueous solutions quickly, accurately, and economically with Orion™ Sodium Electrodes
A unique redox internal system with an Orion™ ROSS™ electrode provides faster response, greater stability, and better accuracy than conventional sodium electrodes.