Corning® BioCoat™ Control Cell Culture Inserts, PET Membrane, Corning
Catalog #: CA-CB354572
Supplier: Corning
Specifications
- Pore size:0.4 µm
- Description:Control cell culture inserts in four 24-well companion plates
- Cat. no.:CA-CB354572
- Membrane:PET
- Supplier No.:354572
Specifications
About this item
Corning BioCoat cell culture inserts are pre-coated with extracellular matrix proteins for applications requiring a protein-coated cell surface, such as cell adhesion, growth, invasion, migration and/or differentiation.
Coatings include Corning Matrigel® matrix, fibronectin, collagen and laminin.
Customers Who Bought This Also Bought