ContactUsLinkComponent
OSOM ULTRA STREP-A 25TST/PK
OSOM® Ultra Strep A Test Kits, Sekisui Diagnostics
Catalog #: CA-71001-042
CAS Number:  
OSOM ULTRA STREP-A 25TST/PK
OSOM® Ultra Strep A Test Kits, Sekisui Diagnostics
Catalog #: CA-71001-042
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    Ultra Strep A Test Kits
  • Cat. no.:
    CA-71001-042
  • Supplier No.:
    147
  • No. of Tests:
    25
Specifications
About this item
These convenient test kits provide easy-to-read, two-color results in as little as five minutes
Studies have shown that the sensitivity of Ultra Strep A is statistically equivalent to single swab culture. Kit contents maintain a room temperature shelf life of 15 months from the date of manufacture.
Certifications: CLIA-waived.
Ordering information: The 50-test kit includes two additional tests for external QC testing.
Customers Who Bought This Also Bought