VWR® Autoclavable PP Bags
Catalog #: 95042-552
Supplier: VWR International
Small Business Enterprise
Specifications
- W×L:254×305 mm (10×12")
- Color:transparent
- Cat. no.:95042-552
Specifications
About this item
Transparent 2 mm thick polypropylene bags are suitable for autoclaving hazardous and nonhazardous materials and waste.
- Can withstand autoclaving temperatures up to 135 °C (275 °F)
Twist ties (included) allow proper closure for bag expansion and venting during sterilization.
Certifications: Meet strength requirements in ASTM D 1709-04 for 165g dart drop test.
Customers Who Bought This Also Bought