VWR® Glove Liners
Catalog #: 94000-610
Supplier: VWR International
Product available on GSA Advantage®
Specifications
- Ambidextrous/Hand-specific:Ambidextrous
- Glove Color:White
- Glove size:M
- Description:Full finger liners, with orange cuff
- Disposable / reusable:Reusable
Specifications
About this item
Ergonomically designed glove liners wick away moisture for comfortable use with disposable gloves.
Ergonomically designed glove liners are made from 15 denier, low-lint nylon knit fabric. Liners provide protection and comfort when used with latex, nitrile, and PVC gloves. Ambidextrous. Fully launderable for reusability and economy. Color-coded cuffs indicate size.
- Ambidextrous glove liners
- Reduces perspiration and wicks away moisture
- Low-lint, 100% 15 denier nylon composition
- Sturdy, launderable, and reusable
- Color-coded cuffs indicate size
- Pre-laundered; will not shrink
Liners provide protection and comfort when used with latex, nitrile, and PVC gloves. Fully launderable for reusability and economy.
Packaging: Glove Liners are packaged 20 pair per bag, 15 bags per case. They are packed in clean polyethylene bags that are free of silicone, dioctyl phthalate (DOP), and amides.
Customers Who Bought This Also Bought