VWR® Sample Library Tubes and Closures, 1.2 ml
Catalog #: 93000-042
Supplier: VWR International
Specifications
- Description:8-Well Tube Strips in 8 x 12 Racks, Sterile
- Environmentally Preferable:Sustainable Packaging,Low Manufacturing Impact
- Cat. no.:93000-042
Specifications
About this item
These 1.2 ml Library and Cluster Tubes are ideal for storage, dilution, mixing, harvesting, culture assays and screening based assays.
- SBS footprint; 8×12 format to fit automated platforms
- Available as individual tubes, strips of 8 tubes, and strips of 12 tubes
- Tubes are made of virgin polypropylene and caps are made of polyethylene to provide a safe and secure seal
- Certified free of detectable DNase, RNase, DNA, and PCR inhibitors
- Sterile configurations are also pyrogren-free
Racks are printed with an alphanumeric grid, meet SBS standards, and can be used in robotic applications. Polypropylene racks, tubes, and lid are autoclavable and freezable to –90 °C (–130 °F). Polyethylene closure strips are also available seperately.
Ordering information: Racked tubes are supplied with a rack lid. Tube closure strips must be purchased seperately.
Customers Who Bought This Also Bought