Specifications
- Volume:300 µl
- Environmentally Preferable:Sustainable Packaging,Safer Human and Environmental Health
- Tip Color:Clear
- Packed:96/Rack
- Tip Style:Universal Fit Tips, Low Retention
- Supplier No.:2092
Specifications
About this item
The exclusive S³ polymer delivers to within 0.1% of indicated volume, eliminating sample hold-up and ensuring a more precise application.
Eco-friendly empty pipet tip trays are manufactured from recycled polypropylene. The trays are autoclavable, enabling reuse for maximum sustainability. When combined with ESP™ reload systems, these tip trays reduce plastic waste by up to 90%. The trays provide superior stability, firm placement, and a maximum space saving design.
- Autoclavable
- Good sustainability
- Superior stability, firm placement
- Maximum space saving design
The exclusive S³ polymer delivers to within 0.1% of indicated volume, eliminating sample hold-up and ensuring a more precise application.
Eco-friendly empty pipette tips trays are manufactured from recycled polypropylene. When combined with ESP™ reload systems, these tip trays reduce plastic waste by up to 90%.
This proprietary technology is essential for quantitative assays where sample retention can hinder accurate results. Universal pipette tips are made from virgin polypropylene and are equipped with a thin wall design that provides an optimal seal with industry leading pipettors.