uTIP™ Filtered Pipette Tips for Universal Pipettors, Biotix
Catalog #: 89136-572
Supplier: Biotix
Product available on GSA Advantage®
Specifications
- Volume:10 µl
- Packed:96 Tips/Rack, 10 Racks/Pack, 5 Packs/Case
- Filter:+
- Sterile:+
- Tip Style:uTIP™ Filtered, Low Retention
- Supplier No.:M-0010-9FC
Specifications
About this item
Biotix filtered tips provide protection against aerosol contamination (cross contamination) with a 99.99% bacterial filter efficiency (BFE). uTIP is compatible with major brands like Eppendorf, Gilson, Biohit and Thermofisher.
- FlexFit® technology features a flexible proximal end to ensure a secure seal with less force then competitor tips
- X-Resin® is our proprietary low retention resin which maximizes sample recovery and improves accuracy and precision
- Blade® Technology makes a fine point at the distal end of the tip ensuring a full dispense without the need for tip touch off
Each lot is tested for accuracy and precision, ensuring superior quality and compatibility. All raw materials are controlled and inspected to ensure only the best go into our products. All racks include a label with a unique bar-coded serial number, part number, and description for traceability.
Customers Who Bought This Also Bought