Specifications
- Cap Size:38-430
- Capacity:1 L (33.8 oz.)
- Height:213 mm
- Neck O.D.:34.5 mm
- Outer Diameter:92 mm
Specifications
About this item
These sterile media bottles are manufactured from Eastar™ PETG, the resin of choice for single use labware.
- PETG resin construction
- Sterile
- Graduated markings
- Sterilized via gamma radiation
Ideal for use in stem cell, mammalian, and in-vitro fertility culture applications, PETG media bottles are designed to retain their clarity after gamma sterilization and consistently maintain the integrity of contents.
Eastar™ PETG resin meets ISO 10993 and/or USP Class VI biocompatibility requirement; Food Contact Status compliant. It is an amorphous material that displays performance properties including clarity, toughness, excellent gas barrier and chemical inertness. Bottles can be safely sterilized with gamma radiation without property loss or color shift. They are supplied gamma sterilized to SAL 10⁻⁶, with purple caps attached.
Bottles are also ideal for the storage of media, laboratory reagents, and biological buffers. They feature a patented, leak-proof, tamper-evident closure system. Graduation markings are molded in for reference filling.