Order Entry
ContactUsLinkComponent
15mL Freezer Box
PolarSafe™ Polypropylene Storage Boxes, 15 and 50 ml, Argos Technologies
Catalog #: 89128-208
Supplier:  Argos Technologies
Small Business Enterprise
15mL Freezer Box
PolarSafe™ Polypropylene Storage Boxes, 15 and 50 ml, Argos Technologies
Catalog #: 89128-208
Supplier:  Argos Technologies
Small Business Enterprise

Specifications

  • Color:
    Blue
  • Description:
    15ml Polypropylene Storage Box
  • Cat. no.:
    89128-208
  • Supplier No.:
    R0015

Specifications

About this item

Polypropylene freezer boxes accommodate 15 & 50mL centrifuge tubes for long term storage; an excellent alternative to cardboard storage boxes

Boxes can be stored to -90°C (130°F) and are autoclavable to 122°C (251.6°F). Accommodate most 4" stainless steel freezer racks.

Clear lid and easy-to-read numerical grid allows for tube reference and identification. White top portion allows for easy labeling or marking.

Customers Who Bought This Also Bought