Order Entry
ContactUsLinkComponent
VWR PETG Square Media Bottles
VWR® Media Bottles, Square, PETG
Catalog #: 89095-296
Supplier:  VWR International
VWR PETG Square Media Bottles
VWR® Media Bottles, Square, PETG
Catalog #: 89095-296
Supplier:  VWR International

Specifications

  • Cap Size:
    38-430
  • Capacity:
    250 ml (8.5 oz.)
  • Height:
    141 mm
  • Cat. no.:
    89095-296
  • Neck O.D.:
    34.5 mm
  • Outer Diameter:
    61 mm

Specifications

About this item

These sterile media bottles are manufactured from Eastar™ PETG, the resin of choice for single use labware.

  • PETG resin construction
  • Sterile
  • Graduated markings
  • Sterilized via gamma radiation

Ideal for use in stem cell, mammalian, and in-vitro fertility culture applications, PETG media bottles are designed to retain their clarity after gamma sterilization and consistently maintain the integrity of contents.

Eastar™ PETG resin meets ISO 10993 and/or USP Class VI biocompatibility requirement; Food Contact Status compliant. It is an amorphous material that displays performance properties including clarity, toughness, excellent gas barrier and chemical inertness. Bottles can be safely sterilized with gamma radiation without property loss or color shift. They are supplied gamma sterilized to SAL 10⁻⁶, with purple caps attached.

Bottles are also ideal for the storage of media, laboratory reagents, and biological buffers. They feature a patented, leak-proof, tamper-evident closure system. Graduation markings are molded in for reference filling.

Ordering information: Each case is specifically packaged for sterile production with multi-barrier elastomer packaging.
Customers Who Bought This Also Bought