Order Entry
ContactUsLinkComponent
RACK SLANTER
SP Bel-Art Rack Slanter, Bel-Art Products, a part of SP
Catalog #: 89093-458
RACK SLANTER
SP Bel-Art Rack Slanter, Bel-Art Products, a part of SP
Catalog #: 89093-458

Specifications

  • Description:
    Rack Slanter
  • Cat. no.:
    89093-458
  • Supplier No.:
    188900100

Specifications

About this item

Tilt test tube racks to 25°, 35°, or 45°.

  • Adjustable angle
  • Ideal for creating agar slant tubes
  • Built-in magnifier aids in viewing Petri dishes
  • Autoclavable

Adjusting the angle of racks reduces hand strain when working with pipettors

Base legs have predetermined slots that the stand slides in and out of to easily change angles. Fits racks up to 29.3×24.2×8.3 cm (119/16×99/16×31/4"). Steam autoclavable at 121 °C (250 °F).

Also ideal for creating agar slant tubes or when using the Mini-Magnifier (54848-306) to aid in the viewing of Petri dishes.

Customers Who Bought This Also Bought