BRAND PCR 8- and 12-Strip Tubes and Caps, BrandTech®
Catalog #: 89093-042
Supplier: Brandtech
Specifications
- Cap Type:Flat (qPCR)
- Capacity:0.2 ml
- Format:Strip
- Color:Clear
- Description:8-Tube Strips with Individual Attached Caps
- Cat. no.:89093-042
- Supplier No.:781332
Specifications
About this item
BRAND PCR tube strips and strip caps are manufactured from high quality virgin resins without lubricants or chemical additives for reproducible results in PCR applications.
- Tubes are autoclavable
- Supplied RNase-, DNase- and DNA-free
New Rigid strips with attached caps for easy handling. Flexible strips for easy strip division.
Strip tubes without attached caps have a tab on the end for easy handling.
Customers Who Bought This Also Bought