Order Entry
ContactUsLinkComponent
PLATE SPOT 9 HOLE PK=6
PYREX® 9 Depression Spot Plate, Corning
Catalog #: 89090-482
Supplier:  Corning
Product available on GSA Advantage®
PLATE SPOT 9 HOLE PK=6
PYREX® 9 Depression Spot Plate, Corning
Catalog #: 89090-482
Supplier:  Corning
Product available on GSA Advantage®

Specifications

  • Description:
    9 Depression Glass Spot Plates
  • Cat. no.:
    89090-482
  • Supplier No.:
    7220-85

Specifications

About this item

These pressed PYREX® glass plates have 9 concave depressions which is 7 mm deep and has 22 mm outer diameter.

The plate is made of borosilicate glass which has size 85×100 mm.

Customers Who Bought This Also Bought