Order Entry
ContactUsLinkComponent
CYLNDR GRAD TC CLSA 250ML PK1
PYREX® VISTA™ Graduated Cylinders, Class A, To Contain, Corning
Catalog #: 89090-452
Supplier:  Corning
   Low Manufacturing Impact
Product available on GSA Advantage®
CYLNDR GRAD TC CLSA 250ML PK1
PYREX® VISTA™ Graduated Cylinders, Class A, To Contain, Corning
Catalog #: 89090-452
Supplier:  Corning
   Low Manufacturing Impact
Product available on GSA Advantage®

Specifications

  • Capacity:
    250 ml
  • Environmentally Preferable:
    Low Manufacturing Impact
  • Environmental certification:
    I-RECs, ISO 14001 - Monterrey
  • Form:
    Tall
  • Cat. no.:
    89090-452
  • Supplier No.:
    70022-250

Specifications

About this item

PYREX® borosilicate glass, class A.

  • Calibrated to contain (TC) in accordance with ASTM E-1272
  • Hexagonal base with graduations in blue enamel
  • 10 ml cylinder has a funnel top
Customers Who Bought This Also Bought