Accessories for SP Bel-Art Clavies®, Biohazard Bag Holders, Autoclavable, Bel-Art Products, a part of SP
Catalog #: 89089-084
Supplier: Bel-Art Products, a Part of SP
Specifications
- Description:Bag holder cover, fits medium or large Clavies® biohazard bag holders, 315×315 mm (12.5×12.5")
- Cat. no.:89089-084
- Supplier No.:131920102
Specifications
About this item
Customers Who Bought This Also Bought