Order Entry
ContactUsLinkComponent
BEAKER PP GRIFFIN 100ML PK12
Beakers, Griffin Low Form, Ace Glass Incorporated
Catalog #: 89085-372
Supplier:  Ace Glass
Product available on GSA Advantage®
Small Business Enterprise
BEAKER PP GRIFFIN 100ML PK12
Beakers, Griffin Low Form, Ace Glass Incorporated
Catalog #: 89085-372
Supplier:  Ace Glass
Product available on GSA Advantage®
Small Business Enterprise

Specifications

  • Capacity:
    100 ml (3.4 oz.)
  • Description:
    Griffin Low Form Beaker
  • Cat. no.:
    89085-372
  • Supplier No.:
    12420-08

Specifications

About this item

These Griffin-style polypropylene beakers are for general laboratory use

  • Autoclavable
  • Approximate raised volume scales
  • Tapered walls for safe handling and convenient stacking

Combines a “no-drip” pouring spout with unbreakability and maximum translucency.

Ordering information: Lids are not included.
Customers Who Bought This Also Bought