PCR Plate Sealers, Sorenson BioScience
Catalog #: 89085-360
Supplier: Sorenson Bioscience
Specifications
- Sterility:Non sterile
- Description:Clear Seal Plate
- Supplier No.:36190
Specifications
About this item
Adhesive seals and cap strips prevent evaporation during thermal cycling or storage and are compatible with Sorenson BioScience PCR plates and tube strips.
Certified free of RNase / DNase, Human DNA, Pyrogens and PCR Inhibitors.
Customers Who Bought This Also Bought