Axygen® 24-Well Deep Well Plates, Rectangular Wells, Corning
Catalog #: 89080-534
Supplier: Corning
Product available on GSA Advantage®
Specifications
- Sterility:Sterile
- Description:Deep Well Plates
- Environmentally Preferable:Low Manufacturing Impact
- Environmental certification:I-RECs, ISO 14001 - Reynosa
- Working Well Volume:10 ml
- Supplier No.:P-DW10ML24CS
Specifications
About this item
Axygen® deep well and assay plates are used for sample collection, in vitro growth chambers and long-term storage
Made of polypropylene, they offer excellent chemical resistance and will withstand temperatures down to -80°C. Ultra-flat surface ensures proper sealing with heat sealing film. Clear, V Shaped Well Bottom.
Customers Who Bought This Also Bought