Axygen® Tip Refill System, Empty Racks, Corning
Catalog #: 89080-486
Supplier: Corning
Product available on GSA Advantage®
Specifications
- Description:Empty racks; 10 racks/pack, 3 packs/case
- Cat. no.:89080-486
- For Use With:1000, 1200 µl Axygen® tips
- Supplier No.:RFL-R-1200
Specifications
About this item
Axygen® Tip Refill System contains durable refill rack that can be easily refilled by hand and is autoclavable.
- RFL-R-1200 is suitable for 1200 µl refill inserts
- RFL-R is suitable for 10 and 200 µl refill inserts
- RNase- and DNase-free, non pyrogenic
- Translucent rack allows tips to be easily seen
- Color-coded insert allows for easy identification of tip
They are available in different colors and are packaged in bulk or racks.
Customers Who Bought This Also Bought