Order Entry
ContactUsLinkComponent
Heavy Duty Maintenance Wipers
PIG® Heavy-Duty Maintenance Wipers, New Pig
Catalog #: 89069-042
Supplier:  New Pig
Heavy Duty Maintenance Wipers
PIG® Heavy-Duty Maintenance Wipers, New Pig
Catalog #: 89069-042
Supplier:  New Pig

Specifications

  • W×L:
    30.5 x 33.7cm (12 x 13.25")
  • Color:
    White
  • Cat. no.:
    89069-042
  • Packaging:
    1/4 -fold
  • Weight:
  • Supplier No.:
    WIP303

Specifications

About this item

Strong and solvent-resistant for wiping machines and rough parts, but soft enough for cleaning skin or preparing surfaces.

  • Excellent for heavy-duty maintenance jobs like machine wipedowns and degreasing parts
  • Low-lint construction won't shed, pill, or leave behind unwanted fibers when wiping parts
  • Binder-free construction withstands solvents, degreasers, and cleaning agents

Wipers are 1/4 folded and poly wrapped for easy handling and loading. Convenient pop-up box lets you take just one wiper at a time while keeping remaining stock clean and dry. Pop-up Z-box keeps wipers clean, neat and organized for quick access. Economical jumbo rolls dispense easily and offer a lower cost per wiper for high-volume use.

Exceptional abrasion resistance and wet strength that stands up to rough surfaces, jagged edges or metal chips.

Customers Who Bought This Also Bought