Order Entry
ContactUsLinkComponent
Full Face Shield With Head Strap
Face Shield, Unimed-Midwest
Catalog #: 89068-863
Supplier:  Unimed-Midwest
Full Face Shield With Head Strap
Face Shield, Unimed-Midwest
Catalog #: 89068-863
Supplier:  Unimed-Midwest

Specifications

  • Description:
    Full Face Shield
  • Size:
    13"W x 7.5" L
  • Cat. no.:
    89068-863
  • Supplier No.:
    SSDS00205B

Specifications

About this item

Splash shield provides comfortable face protection.

  • Soft elastic strap wraps around head to secure shield
  • Anti-fog
  • Optically Clear
  • 13 inch impenetrable foam barrier for wide coverage
  • Disposable and single use

It is not recommended to disinfect and reuse shields because doing so would destroy the anti-fog coating.

Customers Who Bought This Also Bought