Multi-Well Reagent Reservoirs
Catalog #: 89049-030
Supplier: Excel Scientific
Specifications
- Capacity:175 ml
- Sterility:Non sterile
- Description:Reservoir with Lid
- Supplier No.:RSVL175N10
Specifications
About this item
Reusable, large-capacity reservoirs feature a V-shaped basin that is compatible with both eight- and twelve-channel pipettors
- Available with or without lids
- Constructed of clear, chemical-resistant polypropylene for constant sample visibility
- Molded graduation marks provide volume indication at 25, 50, 75, 100, 125, and 150mL
- Autoclavable
The removable lids latch tightly for secure storage, and offer simple access that can be manipulated with one hand. Reservoirs are stackable with lids closed, and a perimeter ridge on each lid prevents sliding. A reinforced base eliminates wobbling and the threat of spilling.
They provide an environmentally sound alternative to disposable reservoirs, allow for fewer refills, and ensure minimal sample waste.
Customers Who Bought This Also Bought