VWR® Signature™ Disposable Electroporation Cuvettes
Catalog #: 89047-208
Supplier: VWR International
Product available on GSA Advantage®
Specifications
- Volume:400 µl
- Description:Round Cap
- Gap Size:2 mm
Specifications
About this item
VWR Signature™ Disposable Electroporation Cuvettes are designed for use with bacterial, yeast, fungi, and mammalian cells.
- Cuvettes are precision built to provide exacting tolerances, ensuring reproducible results
- Polished aluminum plates reduce static and arcing
- Compatible with most electroporation devices
- Gamma irradiated for sterility
Disposable cuvettes are manufactured from high quality, durable polycarbonate material that has been tested to optimize transformation efficiencies.
Ordering information: Each pack contains 50 individually wrapped cuvettes.
Customers Who Bought This Also Bought