Microcentrifuge Tubes with Screw Cap, Molecular BioProducts
Catalog #: 89047-094
Supplier: Thermo Scientific
Specifications
- Volume:2 ml
- Style:Conical-Bottom
- Supplier No.:3468
Specifications
About this item
Manufactured from high quality polypropylene, these clear microcentrifuge tubes are designed for vapor phase liquid nitrogen storage, boiling, and autoclaving procedures.
- RNase-, DNase-, and endotoxin-free
- Nonsterile
Tubes are sturdy enough to withstand centrifugation at 20000×g. All tubes are supplied with unattached screw cap and O-ring. Conical bottom tubes allow for more complete sample recovery. Freestanding tubes feature external ribs that assist in positioning and locking tubes into racks, allowing for convenient one-handed opening and closing.
Customers Who Bought This Also Bought