Pipettor Reagent Reservoirs, DWK Life Sciences
Catalog #: 89044-716
Supplier: DWK Life Sciences (KIMBLE)
Small Business Enterprise
Specifications
- Capacity:48 ml
- Length:12.6 cm (5")
- Sterility:Sterile
- Width:8.3 cm (3¼")
- Description:12-channel reservoir
- Material:PS
- Supplier No.:W851368
Specifications
About this item
Reagent reservoirs are designed for maximum recovery of liquid reagents.
- Available in PP or PS
- Sterile polystyrene reagent reservoirs are available in 8- or 12-channel formats
V-shaped 75 ml polypropylene reservoirs with open wells are well-suited for single-and multi-channel pipetting. The 125 ml reservoir includes a hinged lid.
Customers Who Bought This Also Bought