Order Entry
ContactUsLinkComponent
VWR Pestle
VWR® Mortars and Pestles, Porcelain
Catalog #: 89038-160
Supplier:  VWR International
VWR Pestle
VWR® Mortars and Pestles, Porcelain
Catalog #: 89038-160
Supplier:  VWR International

Specifications

  • Length:
    114 mm
  • Description:
    for Mortar 47 x 70 mm
  • Cat. no.:
    89038-160

Specifications

About this item

Mortars are constructed of chemical porcelain and feature a lip and a glazed exterior.

  • Porcelain construction
  • Glazed exterior, unglazed interior
  • Oversized pestle for comfort

Pestles are constructed entirely of chemical porcelain or with a porcelain grinding head and wooden handle. Wooden handle is not autoclavable. Pestles are also glazed to grinding surface and oversized for hand comfort.

Customers Who Bought This Also Bought