Order Entry
ContactUsLinkComponent
VWR Dessicator Plate
VWR® Desiccator Plates
Catalog #: 89038-074
Supplier:  VWR International
VWR Dessicator Plate
VWR® Desiccator Plates
Catalog #: 89038-074
Supplier:  VWR International

Specifications

  • Ø:
    230 mm
  • Hole Diameter:
    2 mm
  • Cat. no.:
    89038-074

Specifications

About this item

Chemical-porcelain dessicator plates are perforated with numerous small holes and a large center hole.

  • Autoclavable
  • Plates without feet

Plates are also glazed on one side and highly resistant to chemicals. For use in temperatures up to 1150 °C (2102 °F).

Customers Who Bought This Also Bought