Specifications
- Description:Activated charcoal filter VaporGuard
- Cat. no.:89012-608
- Supplier No.:931401
Specifications
About this item
The scavenging interface kit features an interface valve that is installed between an anesthesia machine and a vacuum evacuation system. The kit helps control and monitor exposure to anesthetic waste gases.
- Valve equalizes both positive and negative pressure so that no metabolic gases are pulled past the patient
- Monitoring badge adsorbs halogenated hydrocarbons, including isoflurane and halothene
- After <=8 hours of exposure, the badge should be sent to a chemical laboratory for analysis of exposure levels
The activated charcoal adsorption filter adsorbs passively and has been proven effective in eliminating waste anesthetic gas pollution.