Axygen® Mini Tube Systems (MTS) Caps, Corning
Catalog #: 89005-726
Supplier: Corning
Product available on GSA Advantage®
Specifications
- Sterility:Non sterile
- Description:Replacement Caps for 8-Tube Strip
- Supplier No.:MTS-8CP-C
Specifications
About this item
Axygen® mini tube systems are available in 8-cap or 12-cap strip format.
- Molded-in condensation rings on the lid minimize cross contamination
Customers Who Bought This Also Bought