Order Entry
ContactUsLinkComponent
VWR* Good Lab Practice Balance Calibration Log Book
VWR® Good Laboratory Practice Log Books
Catalog #: 89005-184
Supplier:  VWR International
VWR* Good Lab Practice Balance Calibration Log Book
VWR® Good Laboratory Practice Log Books
Catalog #: 89005-184
Supplier:  VWR International

Specifications

  • Cover Color:
    Hunter green
  • Description:
    Balance calibration log book
  • Cat. no.:
    89005-184
  • Length:
    287 mm (11.7")
  • Width:
    225 mm (8.9")

Specifications

About this item

Log books feature user data, guidelines, and professionally designed documentation pages for the intended purpose.

  • Designed to lay flat
  • Flexible imitation leather cover

Acid-free, archival-safe pages are sequentially numbered. Case is bound in a flexible imitation leather cover specially coated for durability in laboratory environments.. Equipment log books have 96 pages; all other log books have 168 pages.

Customers Who Bought This Also Bought