ContactUsLinkComponent
Burets Threaded Closure Fit Straight Bore PTFE Stopcock with Dust Cap
Accessories for KIMAX® Burets with Threaded Closure Fit, Straight Bore PTFE Stopcock, Kimble Chase
Catalog #: 89002-166
Burets Threaded Closure Fit Straight Bore PTFE Stopcock with Dust Cap
Accessories for KIMAX® Burets with Threaded Closure Fit, Straight Bore PTFE Stopcock, Kimble Chase
Catalog #: 89002-166
Specifications
  • Description:
    PTFE LUBRI-FLO® stopcock, 2.8 mm
  • Cat. no.:
    89002-166
  • Supplier No.:
    41007F-2
  • Details:
    For use with 29053H and 17121H