Order Entry
ContactUsLinkComponent
KIMAX* Reusable Chemical Test Tube
KIMAX® Test Tubes, Plain, Kimble Chase
Catalog #: 89001-448
KIMAX* Reusable Chemical Test Tube
KIMAX® Test Tubes, Plain, Kimble Chase
Catalog #: 89001-448

Specifications

  • Capacity:
    30 ml
  • Ø:
    19 mm
  • Height:
    150 mm
  • Supplier No.:
    45050 19150

Specifications

About this item

KIMAX-51® tubes are constructed from N-51A borosilicate glass tubing and feature a round bottom, sturdy lip, and uniform wall thickness for maximum heat transfer and chemical resistance.

Tubes are supplied without a marking spot or cap.

Customers Who Bought This Also Bought