Order Entry
ContactUsLinkComponent
KIMAX* brand Erlenmeyer Flasks, Wide Mouth, Heavy-Duty Tooled Top Finish
KIMAX® Erlenmeyer Flasks, Wide Mouth, Heavy-Duty Tooled Top, Graduated, Kimble Chase
Catalog #: 89001-358
KIMAX* brand Erlenmeyer Flasks, Wide Mouth, Heavy-Duty Tooled Top Finish
KIMAX® Erlenmeyer Flasks, Wide Mouth, Heavy-Duty Tooled Top, Graduated, Kimble Chase
Catalog #: 89001-358

Specifications

  • Capacity:
    1000 ml
  • Graduation Interval:
    50 ml
  • Graduation range:
    250 - 1000 ml
  • Cat. no.:
    89001-358
  • Stopper No.:
    11
  • Supplier No.:
    26650-1000

Specifications

About this item

Flasks feature a wide mouth, thick walls, and tapered contours to minimize chipping.

All flasks have white scales to indicate approximate volumes. Designed from ASTM Specification E1404, Type I, Class 2 requirements.

Customers Who Bought This Also Bought