Polyethylene [ST] Stoppers, Kimble Chase
Catalog #: 89000-803
Supplier: DWK Life Sciences (KIMBLE)
Specifications
- Cat. no.:89000-803
- ST Stopper No.:19
- Supplier No.:28160R-19
Specifications
About this item
For items ground to [ST] stopper dimensions
Stoppers are yellow and blue with a closed bottom. Made from linear high density polyethylene. Enlarged flange is designed to protect the neck if the glassware item is tipped over.
Customers Who Bought This Also Bought